Order Entry
Slovakia
ContactUsLinkComponent
Universal sorbent rolls
Universal sorbent rolls
Catalog # BRDY813828
Supplier:  BRADY
Universal sorbent rolls
Catalog # BRDY813828
Supplier:  BRADY
Specifications
  • Description:
    Universal sorbent rolls, heavyweight, perforated and bonded
  • L×W:
    44000×4800 mm
  • Environmentally Preferable:
  • Absorbency:
    192 L
  • Pk:
    2 Roll
Specifications
About this item
These grey coloured medium weight, bonded and perforated universal sorbent rolls absorbs industrial liquids including oils, water, solvents, coolants.
  • Made of one-ply, meltblown, bonded
  • Strong and durable with a superior finish with less lint for improved abrasion resistance
Ideal to cover a large surface area for fast clean up of spills, leaks and drips. Suited for industrial applications where low cost and high absorbency are required.