Supelco® Drug and metabolite standards, JWH-210 5-Hydroxypentyl metabolite solution, (CRM)
S-051-1ML
: Merck
:
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- Description:JWH-210 5-Hydroxypentyl metabolite solution
- Environmentally Preferable:
- Pk Info:ampoule of 1 ml