Paracetamol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
CAS Number:103-90-2
:
Formula:C₈H₉NO₂
MDL Number:MFCD00002328
Boiling Pt:388
Melting Pt:168
Density:1.34 g/cm³ (20 °C)
Molecular Weight:151.16 g/mol
Merck Index:13,00048
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-157-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together