Copper(II) acetate monohydrate 98+%, extra pure
: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate
:6046-93-1
:
:(H₃CCOO)₂Cu·H₂O
:MFCD00149570
:240 °C (760 mmHg)
:199.65 g/mol
:115 °C
:8 II
:13,02651
:205-553-3
:3260
:GHS05
:GHS07
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Blue-green to blue Crystalline powder
- Infrared spectrum:Conforms
- Titration Iodometric:≥98.0 %
- Chloride (Cl):≤30 ppm
- Sulfate (SO4):≤100 ppm
- Iron (Fe):≤20 ppm
- Insoluble matter:≤0.01 %
Frequently Bought Together