Polypropylene glycol, M.W. ∼4,000
: Poly(propylene glycol), Poly(propylene oxide), PPG
:25322-69-4
:
:H(OCH₂CH₂CH₂)nOH
:MFCD00084446
:1025
:Ambient
:500-039-8
- Environmentally Preferable:
- Appearance:Clear light yellow to light green Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4500 to 1.4520 (20°C, 589 nm)
Frequently Bought Together