Fullerene C70, Sigma-Aldrich®
: Merck
: Buckminster-Fullerene C70, [5,6]-Fullerene-C70
:115383-22-7
:
:C₇₀
:MFCD00282904
:1.73 g/cm³ (20 °C)
:840.77 g/mol
:> 280 °C
:4.1 III
:M-Clarity™ quality level = MQ100
:1325
- Environmentally Preferable:
Frequently Bought Together