Order Entry
Slovakia
ContactUsLinkComponent
Anti-KLK11 Rabbit Polyclonal Antibody
Anti-KLK11 Rabbit Polyclonal Antibody
Catalog # PRSI47-139
Supplier:  PROSCI
undefined
Anti-KLK11 Rabbit Polyclonal Antibody
Catalog # PRSI47-139
Supplier:  PROSCI

Specifications

  • Antigen name:
    Kallikrein-related peptidase 11
  • Antigen symbol:
    KLK11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    Kallikrein-11 inactive chain 2|Serine protease 20|KLK11|Kallikrein-11 inactive chain 1|PRSS20|Kallikrein11|Hippostasin|UNQ649/PRO1279|Kallikrein 11|TLSP|hK11|Trypsin-like protease|Kallikrein-11
  • Immunogen:
    Synthetic peptide corresponding to aa residues 21-37 of human NKX3.1 conjugated to KLH at the C-terminus.
  • Tested applications:
    Immunoaffinity Chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

NKX3.1 is a homeobox gene which exhibits prostate- and testis-specific expression. Loss of NKX3.1 expression has been implicated in prostate development and tumorigenesis. NKX3.1 expression is dramatically down-regulated in testicular cancer of germ cell origin. Results suggest that NKX3.1 is essential for normal testis function and that its loss of expression is highly associated with the invasive phenotype of testicular germ cell tumors.

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human