Anti-HCN3 Goat Polyclonal Antibody
PRSI42-620
: PROSCI
- Antigen symbol:HCN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:HCN3 antibody was raised against a 13 amino acid synthetic peptide near the internal region of HCN3.
- Size:100 µg
- Pk:100 µG
HCN3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
HCN3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HCN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat