Order Entry
Slovakia
ContactUsLinkComponent
 
Impurity and API standard, [5-[(dimethylamino)methyl]furan-2-yl]methanol hydrochloride
  PROOMM0086.17
 :  PROMOCHEM
 :  
Impurity and API standard, [5-[(dimethylamino)methyl]furan-2-yl]methanol hydrochloride
  PROOMM0086.17
 :  PROMOCHEM
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    81074-81-9
  • Description:
    [5-[(Dimethylamino)methyl]furan-2-yl]methanol hydrochloride
  • Environmentally Preferable:
  • Formula:
    C₈H₁₃NO₂·ClH
  • Description:
    [5-[(Dimethylamino)methyl]furan-2-yl]methanol Hydrochloride