- Antibody type:Primary
- Antigen name:Alpha-Methylacyl-CoA Racemase
- Antigen symbol:AMACR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:23600
- Antigen synonyms:AMACRD|RM|RACE|CBAS4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
alpha-methylacyl-CoA racemase(AMACR/P504S) is Prostate-specific antigen (PSA) screening for prostate cancer is now widespread in the United States among men of all ages. However PSA has limited specificity because benign disease, including prostatic enlargement and inflammation, can increase PSA levels. Thus, a more specific prostate cancer markers is needed. One such potential marker is AMACR, an enzyme that is involved in peroxisomal beta-oxidation of dietary branched-chain fatty acids. Recent studies have shown that, compared with expression in normal or benign prostate epithelium, AMACR is consistently overexpressed in prostate cancer epithelium, making it a specific marker for cancer cells within the prostate gland. Furthermore, overexpression of AMACR may increase the risk of prostate cancer because its expression is increased in premalignant lesions (prostatic intraepithelial neoplasia).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AMACR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat