Specifications
- Antibody type:Primary
- Antigen name:Lactamase beta 2
- Antigen symbol:LACTB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:51110
- Antigen synonyms:CGI 83|Beta lactamase like protein 2|Lactamase beta 2|Beta-lactamase-like protein 2|LACTB2.|LACB2_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Penicillin refers to any member of beta-lactam antibiotics group. These agents are identified by a beta-lactam ring within their molecular structure. As the most widely used group of antibiotics available, beta-lactams are used for the treatment of bacterial infections usually caused by gram-positive organisms. Beta-lactam antibiotics are bactericidal, functioning to inhibit the synthesis of the peptidoglycan layer of bacterial cell walls. Bacterial penicillin-binding proteins and beta-lactamases constitute a large family of serine proteases that perform essential functions in the synthesis and maintenance of peptidoglycan cell wall. Notably, beta-lactamases cleave beta-lactams, therefore providing the bacteria with resistance to the antibiotic. Homologues of beta-lactamases occur in many species, including human, rat, cow, rabbit, pig, xenopus, zebrafish, and C. elegans. The human homologues, LACTB and LACTB2, are active-site-serine enzymes thought to be involved in metabolism.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: LACTB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat