Order Entry
Slovakia
ContactUsLinkComponent
 
Anti-DEFB1 Mouse monoclonal antibody unconjugated [clone: M11-14b-D10]
  ABCAAB14425-50
 :  ABCAM
Anti-DEFB1 Mouse monoclonal antibody unconjugated [clone: M11-14b-D10]
  ABCAAB14425-50
 :  ABCAM

 

  • Antibody type:
    Primary
  • Antigen name:
    defensin, beta 1
  • Antigen symbol:
    DEFB1
  • Clonality:
    Monoclonal
  • Clone:
    M11-14b-D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DEFB1 (defensin, beta 1)
Clonality: monoclonal
Clone: M11-14b-D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human