Anti-GHRH Rabbit Polyclonal Antibody
ABGEAP7758A
: ABGENT
- Antibody type:Primary
- Antigen name:Growth hormone releasing hormone
- Antigen symbol:GHRH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:GRF|GHRF|INN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GHRH (growth hormone releasing hormone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human