Anti-ERCC3 Rabbit Polyclonal Antibody
Catalog # AVIVARP37963_P050
Supplier: AVIVA SYSTEMS BIOLOGY
Specifications
- Antibody type:Primary
- Antigen name:Excision repair cross-complementation group 3
- Antigen symbol:ERCC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2071
- Antigen synonyms:GTF2H|BTF2|TTD2|XPB|TFIIH|RAD25
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ERCC3 (excision repair cross-complementation group 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: