SPE microelution plates, SOLAµ™
Supplier: Thermo Fisher Scientific
Certificates
About this item
SOLA™μ 96 well SPE micro elution plates are specifically designed to meet the increasingly demanding challenges found within bioanalysis. The micro elution format is uniquely positioned to deliver on bioanalytical needs, but limitations with existing
technological design has prevented significant uptake. The SOLA™μ product eliminates these issues by utilising SOLA technology, which removes the need for frits to retain the sorbent material and, in combination with a macro porous structure, allows for a robust, easily controllable platform for reproducible processing of bioanalytical samples.
- High reproducibility at low elution volumes
- Increased sensitivity (×20 fold due to pre-concentration)
- Processing of low volume samples (25 μl)
- Reduced adsorption and solvation issue associated with large molecules
- 96 well formats (for high throughput automation)
- Consistent flow through characteristics with viscous samples
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together