Tin(IV) oxide
LACH30132CP0G01001
: LACH NER
:
- Pack type:Glass bottle for solids
- Pk:1.000 g
- Seq:3047
- Environmentally Preferable:
: Tin(iv)oxide, Stannic Oxide, Tin dioxide
:
:150.71 g/mol
:242-159-0
:18282-10-5
:O₂Sn
:MFCD00011244
:∼2500 °C (1013 hPa)
:1624.85
:6.95 g/cm³ (20 °C)
:14,08778