Order Entry
Slovenia
ContactUsLinkComponent
 
Anti- Pig Polyclonal Antibody (Unconjugated)
  IMMRIR2415
Anti- Pig Polyclonal Antibody (Unconjugated)
  IMMRIR2415

 

  • Antibody type:
    Control
  • Antigen symbol:
    Ig
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Pig
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    10 mg
  • Pk:
    10 mg

 

 

Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Pig
Isotype:
Reactivity: