Anti-ARAP2 Rabbit Polyclonal Antibody
NOVUNBP2-24620
- Antibody type:Primary
- Antigen name:ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2
- Antigen symbol:ARAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:116984
- Antigen synonyms:CENTD1|PARX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: ARAP2 (ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human