Nessler cylinders
These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc
- Made of specially selected borosilicate glass tubing
- Supplied in matched pairs for uniformity
Frequently Bought Together
These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc