Ethyl succinyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethyl 4-chloro-4-oxobutanoate
CAS Number:14794-31-1
:
Formula:C₆H₉ClO₃
MDL Number:MFCD00000751
Boiling Pt:88…90 °C (11 mmHg)
Density:1.163
Molecular Weight:164.59 g/mol
Flash Pt:84 °C (183 °F)
Storage Temperature:Ambient
EINECS:238-855-9
UN:3265
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together