Adenine hydrochloride, (max. 5% H₂O) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride
CAS Number:2922-28-3
:6055-72-7
Formula:C₅H₅N₅·HCl
MDL Number:MFCD00150864
Molecular Weight:171.59 g/mol
Melting Pt:∼ 290 °C (dec.)
EINECS:220-867-0
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together