Order Entry
Slovenia
ContactUsLinkComponent
Random hexamer primer
Random hexamer primer
  SO142
 :  
undefined
Random hexamer primer
  SO142
 :  
  • Pk:
    1 KIT
  • Application:
  • Description:
    Random hexamer primer 24 µg (100 µM)
  • Environmentally Preferable:
Random hexamer primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Random hexamer primer has been functionally tested in first strand cDNA synthesis.