Oligo(dT)₁₈ primer
Supplier: Thermo Fisher Scientific
Certificates
About this item
Oligo(dT)₁₈ primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)₁₈ primer has been functionally tested in first strand cDNA synthesis.
Specifications
- Application:
- Environmentally Preferable:
Specifications
Frequently Bought Together