(1S)-(-)-Camphanic chloride 98%
: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride
:39637-74-6
:
:C₁₀H₁₃ClO₃
:MFCD00135626
:216.67 g/mol
:67…71 °C
:Refrigerator
:254-552-4
:3261
:8 II
:GHS05
- Environmentally Preferable:
- Appearance:White to beige to grey Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:67 to 71 °C
- Specific optical rotation:-17° to -25° (20 °C, 589 nm) (c=3.5, CHCl3)
- Titration RCOCl:≥97.5 %
Frequently Bought Together