Hand disinfectant gel, Stokosept® protect
Supplier: Deb-STOKO
Certificates
About this item
This alcoholic gel combines skin protection, skincare and disinfectant properties in one. It is tested for hygienic (EN 1500) and surgical (EN 12791) hand disinfection without water.
- Bactericidal, fungicidal as well as virucidal (incl. HBV, HIV)
- Contains a high proportion of a purified hamamelis extract, its astringent effect strengthens the epidermis and helps to prevent the skin from softening when wearing gloves
- Care complex consisting of glycerine/panthenol (provitamin B5) for optimum skin compatibility
- Perfume-free, colourant-free
Active ingredients:
60,7 g Ethanol/100 g product
10,0 g Propan-2-ol/100 g product
Suitable for all who need to wear gloves, particularly in hygiene-sensitive areas, such as the food industry, chemical/pharmaceutical industries and the electronics industry.
Certifications: EN 1500, HACCP.
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together