Safety boots, pull-on, Florian® Europe
Firefighter boots made of 2.5-2.7 mm waterproof leather and Gore-Tex® lining. The rubber profile shell sole with street and terrain profile has a 90-degree heel front with ladder bar profile, high cushioning properties and heat and cold insulation of the sole complex. It is oil and petrol resistant and antistatic.
- Waterproof and breathable thanks to Gore-Tex® lining
- Breathable, moisture is transported to the ventilation openings on the shaft and tongue
- Orthopedic properties ensure pressure relief, cushioning, correction of foot position, body straightening and muscle balance
- Sun reflection effect reduces heating of the upper leather
- Increased impact absorption and isolation in the heel area
- Extra wide and high steel toe cap with scuff cap
- Penetration-resistant midsole
- Fire brigade Pictogram to indicate the protective function against hazards during firefighting
Colour: Black
Shaft height: 30 cm
Frequently Bought Together