Acetic acid glacial ≥99.7% ACS
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118
Melting Pt:17
Density:1.05 g/cm³ (20 °C)
Molecular Weight:60.05 g/mol
Flash Pt:38.5
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 II
REACH:01-2119475328-30
:GHS02
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Acetic Anhydride:<0.01%
- Chloride:≤1 ppm
- Colour (APHA):≤10
- Dilution Test:Pass
- Heavy Metals:≤0.5 ppm
- Iron:≤0.2 ppm
- Purity:≥99.7%
- Residue After Evaporation:≤0.001%
- Substances Reducing Dichromate:Pass
- Substances Reducing Permanganate Test:Pass
- Sulphate:≤1 ppm
- Titrable Free Base:≤0.0004 meq/g
Specification Test Results
Frequently Bought Together