siRNA and miRNA transfection reagent, INTERFERin®
PPLU101000016
:
- Pk:5
- siRNA:Yes
- Description:INTERFERin® siRNA transfection reagent, 5×1 ml
- Seq:05
- Environmentally Preferable:
INTERFERin® is a powerful, ready to use siRNA and miRNA reagent which provides more than 90% silencing efficiency at 1 nM siRNA in a wide variety of cells.
- Great silencing using as little as 1 nM siRNA
- Over 90% gene silencing in a wide variety of cells
- Suitable for transfection miRNA and other oligonucleotides
- Gentle mode of action for more robust data and excellent cell viability
- Compatible with serum and antibiotics
Using low concentration of siRNA avoids off-target effects, and its gentle mode of action ensure more robust data and excellent cell viability. Easy to use thanks to its compatibility with serum and antibiotics, INTERFERin® is also perfectly suited for transfection of miRNA and other oligonucleotides such as pre-miRNA, mimic miRNA, anti-miR, etc. Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every 3 years.
Every batch of INTERFERin® is tested in house in a transfection assay on A549-Luc cells, constitutively expressing the Luciferase gene. The silencing efficiency obtained using 1 nM siRNA and INTERFERin® for each batch is indicated on the Certificate of Analysis.
Every batch of INTERFERin® is tested in house in a transfection assay on A549-Luc cells, constitutively expressing the Luciferase gene. The silencing efficiency obtained using 1 nM siRNA and INTERFERin® for each batch is indicated on the Certificate of Analysis.
: 1 ml of INTERFERin® is sufficient to perform 500 to 1000 transfections in 24-well plates.