Precast agarose gels, RunOne™
Supplier: EMBITEC
Certificates
About this item
RunOne™ precast gels do not require a specific gel apparatus but are optimised for use with the RunOne™ Electrophoresis System. A choice of 100, 104 or 108 well formats make these gels convenient for all screening applications.
- Gel format matches that of standard 96-well PCR plates avoiding confusion over results
- Gel is exposed to buffer on both ends resulting in efficient running and less gel distortion
- UV transmissible gel trays make gels easy to handle and view
- Multi-channel compatible wells allow fast and easy gel loading in multiples of 8 or 12
- Short run times - 96 samples plus markers can run in under 20 minutes
Frequently Bought Together