Urine container
Catalog # 216-4739
Supplier: SELECTA
Specifications
- Version:non sterile
- Capacity:100 ml
- Environmentally Preferable:
- Pk:400
Specifications
About this item
From plastic.
A urine sample can be taken safely and clean from the cup without opening it through the special Tapval® opening using a pipette or the supplied dispenser.
Content: 100 ml.
Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904.