Dextran, M.W. ~ 200,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9004-54-0
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935
EINECS:232-677-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together