Bottle top vacuum filtration systems, BOLA
Supplier: Bohlender
Certificates
About this item
Vacuum filters with multi-stage hose connector with integrated lock screw for connecting vacuum tubing with Int.Ø 6 mm or 8 mm. The filters are produced without plasticisers and have an almost universal chemical resistance.
- Collecting and filling vessel made of PFA
- Filling vessel with lid for protection against contaminations
They do not release any trace elements into the filtrate. Due to the non-adhesive surface, they are easy to clean and can be reused.
Certifications: FDA conform.
Frequently Bought Together