Order Entry
Slovenia
ContactUsLinkComponent
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Substances reducing KMnO4:
    Passes test
  • Conductivity (at filling):
    Max. 1 µS/cm
  • TOC (at filling):
    Max. 400 ppb
  • pH (25°C):
    5.0 - 7.5
  • Evaporation residue:
    Max. 1 ppm
  • Ignition residue:
    Max. 1 ppm
  • Silica:
    Max. 0.1 ppm
  • Cl (Chloride):
    Max. 0.05 ppm
  • F (Fluoride):
    Max. 0.05 ppm
  • NH4 (Ammonium):
    Max. 0.2 ppm
  • NO3 (Nitrate):
    Max. 0.01 ppm
  • PO4 (Phosphate):
    Max. 0.01 ppm
  • SO4 (Sulphate):
    Max. 0.1 ppm
  • Ag (Silver):
    Max. 0.0004 ppm
  • Al (Aluminium):
    Max. 0.002 ppm
  • As (Arsenic):
    Max. 0.002 ppm
  • Au (Gold):
    Max. 0.001 ppm
  • B (Boron):
    Max. 0.05 ppm
  • Ba (Barium):
    Max 0.05 ppm
  • Be (Beryllium):
    Max. 0.002 ppm
  • Bi (Bismuth):
    Max. 0.001 ppm
  • Ca (Calcium):
    Max. 0.05 ppm
  • Cd (Cadmium):
    Max. 0.001 ppm
  • Co (Cobalt):
    Max. 0.001 ppm
  • Cr (Chromium):
    Max. 0.0004 ppm
  • Cu (Copper):
    Max. 0.0004 ppm
  • Fe (Iron):
    Max. 0.001 ppm
  • Ga (Gallium):
    Max. 0.004 ppm
  • Ge (Germanium):
    Max. 0.001 ppm
  • In (Indium):
    Max. 0.001 ppm
  • K (Potassium):
    Max. 0.05 ppm
  • Li (Lithium):
    Max. 0.002 ppm
  • Mg (Magnesium):
    Max. 0.005 ppm
  • Mn (Manganese):
    Max. 0.0004 ppm
  • Mo (Molybdenum):
    Max. 0.002 ppm
  • Na (Sodium):
    Max. 0.1 ppm
  • Ni (Nickel):
    Max. 0.0004 ppm
  • Pb (Lead):
    Max. 0.001 ppm
  • Pd (Palladium):
    Max. 0.008 ppm
  • Pt (Platinum):
    Max. 0.001 ppm
  • Sb (Antimony):
    Max. 0.001 ppm
  • Sn (Tin):
    Max. 0.001 ppm
  • Sr (Strontium):
    Max. 0.0004 ppm
  • Ti (Titanium):
    Max. 0.001 ppm
  • Tl (Thallium):
    Max. 0.00005 ppm
  • V (Vanadium):
    Max. 0.001 ppm
  • Zn (Zinc):
    Max. 0.004 ppm
  • Zr (Zirconium):
    Max. 0.001 ppm
  • ISO Standard Water Grade 3:
    Passes test

Specification Test Results

Frequently Bought Together