Lithium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
Boiling Pt:1382
Melting Pt:614
Density:2.07 g/cm³ (20 °C)
Molecular Weight:42.39 g/mol
Merck Index:13,05550
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together