SP Brij® S2 MBAL, Sigma-Aldrich®
SIAL16006-250G
: Merck
:
- Pack type:Poly bottle
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
: Polyethylene glycol octadecyl ether, Steareth-2, Brij 72, Brij S2, Polyoxyethylene (2) stearyl ether
:
:358.60 g/mol
:500-017-8
:GHS07
:Ambient
:9005-00-9
:C₁₈H₃₇(OCH₂CH₂)₂OH
:MFCD00074177
:0.893 g/mL (25 °C) (lit.)