2-Phenylquinoline 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:612-96-4
:
Formula:C₁₅H₁₁N
MDL Number:MFCD00011568
Boiling Pt:363 °C (760 mmHg)
Molecular Weight:205.26 g/mol
Melting Pt:82…84 °C
EINECS:210-326-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:82 to 84 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together