Order Entry
Slovenia
ContactUsLinkComponent
Tissue processor, TP1020
Tissue processor, TP1020
Catalog # 632-1546
Supplier:  LEICA HISTOLOGY
undefined
Tissue processor, TP1020
Catalog # 632-1546
Supplier:  LEICA HISTOLOGY
Specifications
  • Weight:
    60 kg
  • Description:
    Tissue processor, TP1020 with fume control system, with vacuum
  • Height:
    780 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The TP1020 tissue processor is a semi-enclosed filtration system that combines proven technology and a modern, functionally enhanced design. Gentle specimen processing and maximum safety at all stages of processing are the result of precise engineering design based on proven and precise mechanics in conjunction with a modern user interface. The carousel has twelve stations. Tissue baskets made of metal with varying capacities of up to 100 cassettes. There is the possibility of interrupting an automatic process for reloading or removing cassettes for special applications before the end of a run.
  • Ergonomic control panel with tactile keyboard and foil-protected LCD
  • Infiltration time separately programmable for each station, fast and complete
  • Immediate or delayed start function up to nine days
  • Easy editing and changing of programmes, even during a processing run
  • Audible alarms, error messages and warning codes
  • Advanced safety concept for the user
Delivery: Supplied with 10 aluminum reagent containers (1,8 litres), 10 reagent container carriers, two wax baths (1,8 litres), a standard tissue basket, two sets of replacement fuses, a set of power cords.