2,4,5,6-Tetrachloro-m-xylene
Catalog # EHERC17382500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
:
:243.95 g/mol
EINECS:212-886-8
:GHS07
CAS Number:877-09-8
Formula:C₈H₆Cl₄
Boiling Pt:272
Melting Pt:115
Density:1.703 g/cm³ (25 °C)