Tubing, silicone, HelixMark®
Supplier: Freudenberg Medical Europe GmbH
Certificates
About this item
Silicone, translucent, platinum-cured.
- Can be sterilised by autoclaving or ethylene oxide as well as by gamma radiation within the user's validation cycles and exposure levels
- Manufactured according to GMP in an FDA-registered production facility for medical products
- Compliant with the biocompatibility requirements of USP, Class VI
Biomedical/pharmaceutical grade tubing, manufactured by Nusil platinum-cured material. The special manufacturing process minimises flow degradation over extended periods.
Hardness (Shore durometer A): 50
Density: 1,14
Delivery: Tubing is double shrink-wrapped in crush resistant boxes. Material certification and lot traceability are included.