Order Entry
Slovenia
ContactUsLinkComponent
Test tube holder
Test tube holder
Catalog # BOCH2281
Supplier:  BOCHEM
undefined
Test tube holder
Catalog # BOCH2281
Supplier:  BOCHEM

Specifications

  • Length:
    130 mm
  • Environmentally Preferable:
  • Pk:
    1
  • Spanning area:
    10 - 25 mm

Specifications

About this item

This test tube holder is made of nickel plated steel. For tubes with a diameter from 10 to 25 mm.