Order Entry
Slovenia
ContactUsLinkComponent
Male bed bottle
Male bed bottle
Catalog # KART591
Supplier:  KARTELL
undefined
Male bed bottle
Catalog # KART591
Supplier:  KARTELL
Specifications
  • Capacity:
    1500 ml
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
HDPE, translucent.
  • With cap
  • Good chemical resistance
  • Easy to clean