Blood collection tubes, BD Vacutainer®, BD CPT™
Supplier: BD Medical
Certificates
About this item
The BD CPT™ glass tube provides a single-step, standardised method for the isolation of Peripheral Blood Mononuclear Cells (PBMCs) - lymphocytes and monocytes from whole blood. In a single process step, up to 15 million PBMCs can be isolated within 20 minutes.
- Centrifuging conditions: 1500 to 1800xg for 20 minutes at 18 to 25 °C
- CPT™ tube are CE marked for in vitro diagnostic use
- Glass, clear
The BD CPT™ tube enables:
Improved sample preparation yield and consistency - Standardises process when compared to manual FICOLLTM gradient separations - Improves reproducibility between sample preparations and technical operators
Faster separations
- No need to prepare FICOLLTM gradients
- Decreased processing time with less manipulation of the sample
Safe isolation of cells
- Reduces risk of cellular contamination with the cells enclosed in the sterile BD Vacutainer® tube
Frequently Bought Together