BD® Blunt Needles
Supplier: BD Medical
Certificates
About this item
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
- The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
- The blunt filter is designed for safe ampoule medication preparation
- BD blunt fill needles have female luer fittings, which mate to male luer fittings
- BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
- Natural rubber latex is not part of the material formulation
- Sterile, singe use
Caution: Note: The BD blunt fill and filter needles are not for skin injection.