3-Nitrobenzaldehyde ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-61-6
:
Formula:C₇H₅NO₃
MDL Number:MFCD00007249
Boiling Pt:164 °C (23 mmHg)
Melting Pt:57…59 °C
Density:1.481 g/cm³ (0 °C)
Molecular Weight:151.12 g/mol
Merck Index:12,06684
EINECS:202-772-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together