Chloroacetaldehyde diethyl acetal ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Chloro-2,2-diethoxyethane, 2-Chloro-1,1-diethoxyethane
CAS Number:621-62-5
:
Formula:C₆H₁₃ClO₂
MDL Number:MFCD00000949
Boiling Pt:156…157 °C
Melting Pt:–45 °C
Density:1.018
Molecular Weight:152.62 g/mol
Flash Pt:50 °C (122 °F)
Storage Temperature:Ambient
EINECS:210-695-4
UN:1993
ADR:3 III
:GHS06
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together