3,3-Dimethyl-1-butene 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:558-37-2
:
Formula:C₆H₁₂
MDL Number:MFCD00008853
Boiling Pt:41 °C (760 mmHg)
Melting Pt:–115 °C
Density:0.65 g/cm³
Molecular Weight:84.16 g/mol
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:209-195-9
UN:3295
ADR:3 II
:GHS07
:GHS08
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3750 to 1.3780 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together