2,3-Dimethyl-1-butene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:563-78-0
:
Formula:C₆H₁₂
MDL Number:MFCD00008923
Boiling Pt:56…57 °C
Melting Pt:–157 °C
Density:0.679
Molecular Weight:84.16 g/mol
Flash Pt:–18 °C (– °F)
Storage Temperature:Ambient
EINECS:209-262-2
UN:3295
ADR:3 II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together