4-Bromo-1,1,2-trifluoro-1-butene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10493-44-4
:
Formula:C₄H₄BrF₃
MDL Number:MFCD00039274
Boiling Pt:95…98 °C
Density:1.639
Molecular Weight:188.97 g/mol
Flash Pt:17 °C (62 °F)
Storage Temperature:Ambient
EINECS:234-019-2
UN:1993
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together