n-Pentane, Sigma-Aldrich®
Catalog # SIAL236705-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: Pentane
:
:72.15 g/mol
EINECS:203-692-4
UN:1265
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
ADR:3 II
Merck Index:13,07193
:M-Clarity™ quality level = MQ100