Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Ethyl alcohol
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.7895 g/cm³ (20 °C)
Molecular Weight:46.07 g/mol
Flash Pt:12 °C (closed cup)
EINECS:200-578-6
UN:1170
ADR:3 II
Merck Index:12,03806
:Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119457610-43
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Residue on evaporation:Max. 0.0005 %
- Water:Max. 0.0030 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC83672.230 spec_VWRC83672.230_T spec_VWRC83672.230DP spec_VWRC83672.269 spec_VWRC83672.290
Product specification
More
VWR® Anhydrous Solvents VWR® for rotary evaporation
Product Literature
Frequently Bought Together